Order Entry
Austria
Orders LinkContactUsLinkComponent
Cuvette Washers

Cuvette Washers

Single washer for 100 ml washing solution
Catalog # WITG2.990.111
Supplier:   witeg Labortechnik
Cuvette Washers
Catalog # WITG2.990.111
Supplier:  witeg Labortechnik
Cuvette Washers

Single washer for 100 ml washing solution

Price
108,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Cuvette washers acc. to Swiegot, wash and dry cuvettes, cells, test tubes or small scale volumetric instruments with outer dimensions not larger than approx. 20 mm.

Activate flushing by lightly pressing inverted cell onto silicone gashet, complete with laboratory bottle GL 45, 500 ml, complete autoclavable.

Specifications
  • Description:
    Single washer for 100 ml washing solution
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.