Height Endurance Mountaineer Helmets
Supplier: Portwest
Certificates
About this item
The Height Endurance Mountaineer helmet is the ultimate evolution in the Height Endurance range. This vented, peak-less helmet is designed for a variety of uses, and is ideal for working at height, rescue operations, and climbing activities.
- Specially designed for working at height
- Y-style, four-point chin strap
- Size adjustable headband via wheel ratchet system
- Inner expanded polystyrene foam
- Hi-visibility colour for visibility in daylight
- Light and robust
- Compatible with PS58 helmet visor holder
- Compatible with PW47 and PS47 clip-on ear muff
The inner EPS foam, is comfortable sweatband and wheel ratchet adjustment ensure optimal fit. Hi-visibility colours ensure visibility in daylight. A wide range of accessories are available.
Up to seven years lifetime from manufacturing date (as long as undamaged).
Certifications: EN 397 (impact/penetration/−30 °C). The inner EPS foam is fully certified to the EN 12492 standard.
Frequently Bought Together