Bottles, HDPE, with Thread
Supplier: BERLIN PACKAGING RIXIUS GMBH
Jump to:
About this item
Round bottles made up of robust HDPE plastic, black.
- With retracted labeling field
- Tasteless and odorless
- Ideal for filling chemical-technical substances
Certifications: UN approval for liquids. For high-quality filling goods that require UN approval , ideal for retention patterns.
2 Options Available Below
Product Details & Documents
Round bottles made up of robust HDPE plastic, black.
- With retracted labeling field
- Tasteless and odorless
- Ideal for filling chemical-technical substances
Certifications: UN approval for liquids. For high-quality filling goods that require UN approval , ideal for retention patterns.
Product Family Options
Product Information
- CapacityThreadColourHeightAvailabilityPrice
- RIXI3-0602-0500-45Bottles, round, 50 g weight
- RIXI3-0602-1000-45Bottles, round, 88 g weight