Order Entry
Austria
ContactUsLinkComponent
Human Recombinant Thiamin Pyrophosphokinase 1 (from Cells)
  PRSI92-378
 :  
Human Recombinant Thiamin Pyrophosphokinase 1 (from Cells)
  PRSI92-378
 :  

 

  • Pk:
    0,05 mg
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt, please minimise freeze - thaw cycles
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Enzyme name:
    Thiamin pyrophosphokinase 1
  • Enzyme synonyms:
    Thiamin pyrophosphokinase 1, hTPK1, Placental protein 20, PP20, Thiamine pyrophosphokinase 1, TPK1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28,2 kD
  • Sequence:
    Met1-Ser243
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Biological assays

 

 

Thiamin pyrophosphokinase 1 (TPK1), belongs to the thiamine pyrophosphokinase family.

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

TPK1 exists as a homodimer, and is detected in heart, kidney, testis, small intestine and peripheral blood leukocytes, and at very low levels in a variety of tissues. TPK1 can catalyse the phosphorylation of thiamine to thiamine pyrophosphate. It can also catalyse the phosphorylation of pyrithiamine to pyrithiamine pyrophosphate.