Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant Sulfotransferase 1C2 (from E. coli)
  PRSI92-132
 :  
Human Recombinant Sulfotransferase 1C2 (from E. coli)
  PRSI92-132
 :  

 

  • Pk:
    0,05 mg
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt, please minimise freeze - thaw cycles
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Enzyme name:
    Sulfotransferase 1C2
  • Enzyme synonyms:
    Sulfotransferase 1C2, ST1C2, Sulfotransferase 1C1, SULT1C#1, humSULTC2, SULT1C2, SULT1C1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    36,3 kD
  • Sequence:
    Met1-Leu296
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM Tris, 0,1 M NaCl, pH 8,5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Sulfotransferase 1C2 (SULT1C2) is a cytosolic enzyme member of the Sulfotransferase 1 family.

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

Human SULT1C2 is primarily expressed in the adult stomach, kidney and thyroid gland, and in the fetal kidney and liver. SULT1C2 catalyzes the sulfate conjugation of drugs, xenobiotic compounds, hormones, and neurotransmitters. SULT1C2 may be involved in the activation of carcinogenic hyroxylamines. It shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF).