ContactUsLinkComponent
Gateway Accessory Kit for OPC™ UA Enablement of Bio Safety Cabinets
Catalog # 135-2907
Supplier:  Thermo Scientific
undefined
Gateway Accessory Kit for OPC™ UA Enablement of Bio Safety Cabinets
Catalog # 135-2907
Supplier:  Thermo Scientific
Specifications
  • Description:
    Thermo Scientific OPC UA Gateway kit for bio safety cabinets (available in all countries except for China), power supply 100 - 240 V, USB 2.0 cable USB-A to USB-B connector
  • Pk:
    1
  • Details:
    For use with Herasafe™ 2030i, Maxisafe 2030i, Herasafe™ 2025, MSC-Advantage™, 1300 series A2, and 1500 series A2 biological safety cabinets
Specifications
About this item
Enable Open Platform Communication Unified Architecture (OPC™ UA) integration of Thermo Scientific™ Biological Safety Cabinets using this gateway accessory kit. Benefit from the full potential of incorporating laboratory equipment into your new or existing Distributed Control Systems (DCS), including paperless record keeping, remote monitoring, cloud analytics and more.
  • 21 CFR Part 11-compliance and GMP needs
  • Support audit requirements with enhanced data traceability
  • Real-time remote monitoring, workflow efficiencies, and assets and cost optimisation and regulatory compliance
  • The gateway accessory kit comes complete with a gateway device, USB 2.0 cable USB-A to USB-B connector and 100 to 240 V power supply
The gateway accessory kit is compatible with the following models:
Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinet
Thermo Scientific™ Maxisafe 2030i Biological Safety Cabinet
Thermo Scientific™ Herasafe™ 2025 Biological Safety Cabinet
Thermo Scientific™ MSC-Advantage™ Biological Safety Cabinet
Ordering information: One kit is needed per biological safety cabinet connection.
Delivery: Supplied with gateway device, USB 2.0 cable USB-A to USB-B connector and 100 to 240 V power supply.