Clamps for conical standard ground joints, KECK™, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
POM. These clips made of temperature resistant polyacetal (POM) are easy, safe and can be fitted quickly.
- Good spring properties
- Colour coded by size
- No risk of scratching the glass
- Will not corrode or bend out of shape
Frequently Bought Together