3-(2-Hydroxyethyl)indole, Sigma-Aldrich®
Catalog # SIALT90301-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Merck Index:12,09930
Boiling Pt:358 °C (1013 hPa)
Melting Pt:55…57 °C
:M-Clarity™ quality level = MQ100
:161.2 g/mol
EINECS:208-393-2
CAS Number:526-55-6
Formula:C₁₀H₁₁NO
MDL Number:MFCD00005659