Order Entry
Austria
ContactUsLinkComponent
Rabbit Thrombin (from Plasma)
Rabbit Thrombin (from Plasma)
Supplier:  Merck
Certificates
Rabbit Thrombin (from Plasma)
Supplier:  Merck

Specifications

  • CAS No.:
    9002-04-4
  • Source:
    Plasma
  • Species:
    Rabbit
  • Enzyme name:
    Thrombin

Specifications

Frequently Bought Together