
Safranine O ≥85% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:477-73-6
:
Formula:C₂₀H₁₉ClN₄
MDL Number:MFCD00011759
:350.85 g/mol
Density:1.447 g/cm³ (22 °C)
:M-Clarity™ quality level = MQ200
EINECS:207-518-8
:GHS05
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Nitrogen:13.6 - 16.8 %
- Dye Content:≥ 85 %
- Solubility (Colour):Dark Red to Very Dark Red
- Solubility (Turbidity)
1 mg/mL, H2O:Clear - Wavelength:530 - 534 nm
- E1%
50% EtOH and H2O:1250 - 1650 - Recommended Retest Period:4 Years
Specification Test Results
Frequently Bought Together