Oleyl amine ≥70% (by GC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: cis-9-Octadecenylamine, (9Z)-9-Octadecen-1-amine, (Z)-Octadec-9-en-1-amine
CAS Number:112-90-3
:
Formula:C₁₈H₃₇N
MDL Number:MFCD00066507
:267.5 g/mol
Boiling Pt:364 °C (1013 hPa)
Melting Pt:21 °C
Density:0.813 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:204-015-5
:GHS05
:GHS07
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together