Order Entry
Austria
ContactUsLinkComponent
 
 
 
1-Methoxy-5-methylphenazinium methyl sulphate ≥95%, Sigma-Aldrich®
 
undefined
 
 
1-Methoxy-5-methylphenazinium methyl sulphate ≥95%, Sigma-Aldrich®

 

  • Appearance (Colour):
    Red to Brown
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Red
  • Solubility (Turbidity)
    1 mg/ml, H₂O:
    Clear
  • Proton NMR spectrum:
    Conforms to Structure
  • Nitrogen:
    ≥7.9%

 

Frequently Bought Together