
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride, Sigma-Aldrich®
: Merck
:146-68-9
:
:C₁₉H₁₃ClIN₅O₂
:MFCD00011961
:505.7 g/mol
:227…229 °C
:M-Clarity™ quality level = MQ200
:205-676-2
:GHS08
:Refrigerator
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, TECHNICAL, demineralised
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals