Order Entry
Austria
ContactUsLinkComponent
Sodium deoxycholate ≥97% (by titrimetric analysis), Sigma-Aldrich®
Irritant
Sodium deoxycholate ≥97% (by titrimetric analysis), Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Sodium deoxycholate ≥97% (by titrimetric analysis), Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Appearance (Color) :
    White to Off-White
  • Appearance (Form):
    Powder
  • Solubility (Color):
    Colorless to Light Yellow
  • Solubility (Turbidity) 50 mg/ml, H₂O:
    Clear
  • Loss on Drying:
    ≤5.0%
  • Titration with HClO₄ (Dry basis):
    >97 %

Specification Test Results

Frequently Bought Together