Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2-Methyltetrahydrofuran
Supelco® Organic Reference Standard, 2-Methyltetrahydrofuran
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 2-Methyltetrahydrofuran
Supplier:  Merck

Specifications

  • CAS No.:
    96-47-9
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H302 - H315 - H318
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00005367
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-MeTHF; Tetrahydro-2-methylfuran; Tetrahydrosilvan
  • Molecular weight:
    86,13
  • Density:
    0,86 g/ml at 25 °C (lit.)
  • Flash point:
    –10,0 °C – Closed cup
  • Boiling point:
    78 - 80 °C (lit.)

Specifications

Frequently Bought Together