Order Entry
Austria
ContactUsLinkComponent
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥98% for synthesis, for GC derivatisation, Supelco®
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥98% for synthesis, for GC derivatisation, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥98% for synthesis, for GC derivatisation, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • APPEARANCE (COLOR):
    Colorless to Faint Yellow
  • APPEARANCE (FORM):
    Liquid
  • PURITY (GC AREA %):
    ≥ 98 %
  • PROTON NMR SPECTRUM:
    CONFORMS TO STRUCTURE
  • Residue after Ignition:
    20 ppm maximum

Specification Test Results

Related Information

Frequently Bought Together