Order Entry
Austria
ContactUsLinkComponent
Diammonium tetrachloropalladate, Sigma-Aldrich®
Diammonium tetrachloropalladate, Sigma-Aldrich®
Catalog # SIAL205850-2G
Supplier:  Merck
CAS Number:  
Diammonium tetrachloropalladate, Sigma-Aldrich®
Catalog # SIAL205850-2G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :284.31 g/mol
Density:2.17 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:237-498-6
CAS Number:13820-40-1
Formula:H₈Cl₄N₂Pd
MDL Number:MFCD00010884