Toluidine blue O, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:92-31-9
:
Formula:C₁₅H₁₆ClN₃S
MDL Number:MFCD00011934
:305.83 g/mol
Merck Index:12,09658
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-146-2
Related Information
Frequently Bought Together