Order Entry
Austria
ContactUsLinkComponent
Phenyl chloroformate, Sigma-Aldrich®
Phenyl chloroformate, Sigma-Aldrich®
Catalog # SIAL167525-100G
Supplier:  Merck
CAS Number:  
Phenyl chloroformate, Sigma-Aldrich®
Catalog # SIAL167525-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:187 °C (1013 hPa)
Melting Pt:–28 °C
Density:1.248 g/cm³ (20 °C)
EINECS:217-547-8
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
Flash Pt:80 °C
 :156.57 g/mol
UN:2746
CAS Number:1885-14-9
Formula:C₇H₅ClO₂
MDL Number:MFCD00000637