Order Entry
Austria
ContactUsLinkComponent
α-Bromo-4-nitro-o-cresol, Sigma-Aldrich®
α-Bromo-4-nitro-o-cresol, Sigma-Aldrich®
Catalog # SIAL162906-5G
Supplier:  Merck
CAS Number:  
α-Bromo-4-nitro-o-cresol, Sigma-Aldrich®
Catalog # SIAL162906-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :232.03
Melting Pt:147 °C
Density:>1 g/cm³ (20 °C)
EINECS:212-248-9
UN:3261
ADR:8,II
CAS Number:772-33-8
Formula:C₇H₆BrNO₃
MDL Number:MFCD00007340