1,3-Cycloheptadiene, Sigma-Aldrich®
: Merck
:4054-38-0
:
:C₇H₁₀
:MFCD00004155
:94.16 g/mol
:121 °C (1013 hPa)
:–110,4 °C
:0,868 g/cm³ (25 °C)
:223-762-8
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals