

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, Supelco®
Catalog # 12124-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:199.25 g/mol
Boiling Pt:130 °C (1013 hPa)
Density:1.07 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:246-331-6
:GHS02
:GHS07
CAS Number:24589-78-4
Formula:C₆H₁₂F₃NOSi
MDL Number:MFCD00000411