Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(p-Tolyl)propionic acid
3-(p-Tolyl)propionic acid, Sigma-Aldrich®
3-(p-Tolyl)propionic acid, Sigma-Aldrich®
SIAL118265-10G
:
Merck
:
3-(p-Tolyl)propionic acid, Sigma-Aldrich®
SIAL118265-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
164.2 g/mol
:
M-Clarity™ quality level = MQ100
:
216-132-9
:
1505-50-6
:
C₁₀H₁₂O₂
:
MFCD00016561