



Glutaraldehyde 8% in aqueous solution
Catalog # SCIA16010
Supplier: SCI MUENCHEN
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:100.12 g/mol
Storage Temperature:Refrigerator
EINECS:203-856-5
UN:2927
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025