Safranine O
Catalog # ROTHT129.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:350.85 g/mol
Density:1.447 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:207-518-8
CAS Number:477-73-6
Formula:C₂₀H₁₉ClN₄
MDL Number:MFCD00011759