Human thrombin (from plasma)
20 AU
Catalog # ROCH10602400001
Marketsource
Supplier: ROCHE
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:20 AU
- Cat. No.:ROCH10602400001
- Species:Human
- Source:Plasma
- Enzyme name:Thrombin
- CAS No.:9002-04-4
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



