Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium permanganate
Potassium permanganate 3% in aqueous solution Reag. Ph. Eur. 1070902
Potassium permanganate 3% in aqueous solution Reag. Ph. Eur. 1070902
Potassium permanganate 3% in aqueous solution Reag. Ph. Eur. 1070902
87919.290
:
VWR Chemicals
:
Potassium permanganate 3% in aqueous solution Reag. Ph. Eur. 1070902
87919.290
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
158.03 g/mol
:
Ambient
:
231-760-3
:
3082
:
9,III
:
GHS09
:
7722-64-7
:
KMnO₄
:
MFCD00011364