Order Entry
Austria
ContactUsLinkComponent
2-Chloroisonicotinonitrile
2-Chloroisonicotinonitrile
Catalog # MOLEM16178775
Supplier:  Molekula
CAS Number:  
2-Chloroisonicotinonitrile
Catalog # MOLEM16178775
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-Chloro-4-cyanopyridine, 2-Chloropyridine-4-carbonitrile
 :
 :138.56 g/mol
UN:3439
ADR:6.1,III
CAS Number:33252-30-1
Formula:C₆H₃ClN₂
MDL Number:MFCD00174318
ADR:6.1 III