Order Entry
Austria
ContactUsLinkComponent
Phthalaldehyde
Toxic
Corrosive
Environment
Phthalaldehyde
Catalog # MOLE29247150
Supplier:  Molekula
CAS Number:  
Toxic
Corrosive
Environment
Phthalaldehyde
Catalog # MOLE29247150
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :134.13 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:132 °C
EINECS:211-402-2
UN:2923
ADR:8,II
 :GHS06
 :GHS05
 :GHS09
CAS Number:643-79-8
Formula:C₈H₆O₂
MDL Number:MFCD00003335