Phenyl chloroformate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1885-14-9
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00000637
Boiling Pt:187 °C (1013 hPa)
Melting Pt:–28 °C
Density:1.248 g/cm³ (20 °C)
Flash Pt:80 °C
EINECS:217-547-8
UN:2746
ADR:6.1 (8),II
:M-Clarity™ quality level = MQ200
:156.57 g/mol
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) decomposes,Hydrolysis
- Melting Point:-28 °C
- Molar Mass:156.57 g/mol
- Boiling Point:188 - 189 °C (1013 hPa)
- Vapor Pressure:0.9 hPa (20 °C)
- Flash Point:69 °C
- Refractive Index:1.511 (20 °C, 589 nm)
- Density:1.24 g/cm3 (20 °C)
- Ignition Point:540 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Frequently Bought Together