Order Entry
Austria
ContactUsLinkComponent
Ammonia solution 10%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Corrosive
Irritant
Environment
Ammonia solution 10%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Catalog # 4.80905.9200
CAS Number:  
Corrosive
Irritant
Environment
Ammonia solution 10%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Catalog # 4.80905.9200
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    200 L
  • Seq:
    5549

Specifications

About this item

 :
 :35.05 g/mol
Density:0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650