Nitric acid 60%, Ultrapure for trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Boiling Pt:∼120 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.37 g/cm³ (20 °C)
EINECS:231-714-2
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:2031
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-22 °C
- Boiling Point:120 °C (1013 hPa)
- Density:1.37 g/cm3 (20 °C)
- pH:- 1 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.01518.1000_TDS_EN ProductPage_101518
Product specification
Merck Pharma Segment Brochure Merck Ultrapur and Suprapur® High Purity Acids and Bases
Product Literature
msds_101518
SDS
Frequently Bought Together