Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 0.1 mol/l (0.1 N) in 2-propanol, Titripur® acc. to DIN 51558 part 1, Supelco®
Flammable
Irritant
Hydrochloric acid 0.1 mol/l (0.1 N) in 2-propanol, Titripur® acc. to DIN 51558 part 1, Supelco®
Supplier:  Merck
Certificates
Flammable
Irritant
Hydrochloric acid 0.1 mol/l (0.1 N) in 2-propanol, Titripur® acc. to DIN 51558 part 1, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Flash Point:
    12 °C (2-Propanol)
  • Explosion Limit:
    2 - 13.4 %(V) (2-Propanol)
  • Density:
    0.79 g/cm3 (20 °C)
  • pH:
     - 0.5 (H2O, 20 °C)
  • Ignition Point:
    425 °C (2-Propanol)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information

Frequently Bought Together