

N,O-Bis(trimethylsilyl)trifluoroacetamide silylation reagent, for GC derivatisation
Supplier: MACHEREY-NAGEL
Certificates
About this item
CAS Number:25561-30-2
:
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269
:257.4 g/mol
Boiling Pt:40 °C (12 mm Hg)
Melting Pt:–41 °C
Density:0.96 g/cm³ (20 °C)
Flash Pt:34 °C
EINECS:247-103-9
UN:2924
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together