
Safranine O, dark green powder C.I. 50240
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:477-73-6
:
Formula:C₂₀H₁₉ClN₄
MDL Number:MFCD00011759
:350.85 g/mol
Density:1.447 g/cm³ (22 °C)
EINECS:207-518-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test :Passes
- UV/Vis: l max (50% ethanol):530 ± 5 nm
Specification Test Results
Frequently Bought Together