Diethyl succinate
Catalog # EHERC16985300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:174.2 g/mol
Merck Index:12,09038
Boiling Pt:217 °C (1013 hPa)
Melting Pt:–19 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:80 °C
Storage Temperature:Ambient
EINECS:204-612-0
CAS Number:123-25-1
Formula:C₈H₁₄O₄
MDL Number:MFCD00009208