Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3-Chlorophenoxy)acetic acid
(3-Chlorophenoxy)acetic acid
(3-Chlorophenoxy)acetic acid
(3-Chlorophenoxy)acetic acid
EHERC11479500
:
EHRENSTORFER
:
(3-Chlorophenoxy)acetic acid
EHERC11479500
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
186.59 g/mol
:
308 °C (1013 hPa)
:
308 °C (1013 hPa)
:
110 °C
:
209-616-6
:
588-32-9
:
(C₆H₄Cl)OCH₂COOH
:
MFCD00015850