Order Entry
Austria
ContactUsLinkComponent
3-Chloro-p-anisidine
3-Chloro-p-anisidine
Catalog # EHERC11420000
Supplier:  EHRENSTORFER
CAS Number:  
3-Chloro-p-anisidine
Catalog # EHERC11420000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :157.6 g/mol
Boiling Pt:253 °C (1013 hPa)
Melting Pt:82 °C
EINECS:226-298-4
UN:2233
ADR:6.1,III
CAS Number:5345-54-0
Formula:C₇H₈ClNO
MDL Number:MFCD00007771