


Nitric acid 69,0 - 70,0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS03
:GHS05
:GHS07
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Appearance:Passes Test
- Assay (HNO₃):69.0 - 70.0 %
- Arsenic (As):≤0.010 ppm
- Color (APHA):≤10
- Residue after Ignition:≤4 ppm
- Chloride (Cl):≤0.1 ppm
- Phosphate (PO₄):≤0.20 ppm
- Sulfate (SO₄):≤0.5 ppm
- Trace Impurities - Aluminum (Al):≤100.0 ppb
- Arsenic and Antimony (as As):≤4.0 ppb
- Trace Impurities - Boron (B):≤50 ppb
- Trace Impurities - Cadmium (Cd), For Information Only:= 200.0 ppb
- Trace Impurities - Calcium (Ca):≤200.0 ppb
- Trace Impurities - Chromium (Cr):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤50 ppb
- Trace Impurities - Gold (Au):≤200.0 ppb
- Heavy Metals (as Pb):≤100 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Magnesium (Mg):≤100.0 ppb
- Trace Impurities - Manganese (Mn):≤100.0 ppb
- Trace Impurities - Mercury (Hg):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤50 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Tin (Sn):≤200.0 ppb
- Trace Impurities - Titanium (Ti):≤200.0 ppb
- Trace Impurities - Zinc (Zn):≤200.0 ppb
Specification Test Results
Frequently Bought Together