


Nitric acid ≥70%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 6020.2500PE
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (acidimetric):69.0 - 70.0 %
- Appearance:passes test
- Color (APHA):max. 10
- Residue after Ignition:max. 4 ppm
- Specific Gravity at 60°/60°F:1.416 - 1.420
- Trace Impurities (in ppm) Chloride (Cl):max. 0.1
- Trace Impurities (in ppm)Phosphate (PO4):max. 0.2
- Trace Impurities (in ppm)Sulfate (SO4):max. 0.5
- Arsenic and Antimony (as As):max. 4
- Aluminium (Al):max. 100
- Gold (Au):max. 200
- Boron (B):max. 50
- Calcium (Ca):max. 200
- Chromium (Cr):max. 100
- Copper (Cu):max. 50
- Iron (Fe):max. 100
- Mercury (Hg):max. 5
- Potassium (K):max. 300
- Magnesium (Mg):max. 100
- Manganese (Mn):max. 100
- Nickel (Ni):max. 50
- Lead (Pb):max. 100
- Tin (Sn):max. 200
- Titanium (Ti):max. 200
- Zinc (Zn):max. 200
- Heavy Metals (as Pb):max. 100
Specification Test Results
About this item
:
:63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,I
:GHS03
:GHS05
:GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349