Order Entry
Austria
ContactUsLinkComponent
Nitric acid ≥70%, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Corrosive
Toxic
Nitric acid ≥70%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 6019.9200PE
Supplier:  Avantor
CAS Number:  
Oxidizing
Corrosive
Toxic
Nitric acid ≥70%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 6019.9200PE
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay (acidimetric):
    69.0 - 70.0 %
  • Appearance:
    passes test
  • Color (APHA):
    max. 10
  • Residue after Ignition:
    max. 4 ppm
  • Specific Gravity at 60°/60°F:
    1.416 - 1.420
  • Trace Impurities (in ppm) Chloride (Cl):
    max. 0.1
  • Trace Impurities (in ppm) Phosphate (PO4):
    max. 0.2
  • Trace Impurities (in ppm) Sulfate (SO4):
    max. 0.5
  • Trace Impurities (in ppb) Arsenic and Antimony (as As):
    max. 4
  • Trace Impurities (in ppb) Aluminium (Al):
    max. 100
  • Trace Impurities (in ppb) Gold (Au):
    max. 200
  • Trace Impurities (in ppb) Boron (B):
    max. 50
  • Calcium (Ca):
    max. 200
  • Chromium (Cr):
    max. 100
  • Copper (Cu):
    max. 50
  • Iron (Fe):
    max. 100
  • Mercury (Hg):
    max. 5
  • Potassium (K):
    max. 300
  • Magnesium (Mg):
    max. 100
  • Manganese (Mn):
    max. 100
  • Nickel (Ni):
    max. 50
  • Lead (Pb):
    max. 100
  • Tin (Sn):
    max. 200
  • Titanium (Ti):
    max. 200
  • Zinc (Zn):
    max. 200
  • Heavy Metals (as Pb):
    max. 100

Specification Test Results

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,I
 :GHS03
 :GHS05
 :GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349