0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Trays/Dishes/Pots
Laboratory Trays/Dishes
Trays, Chip
Trays, Chip
ENTEH20N21441166C2
:
ENTEGRIS
Trays, Chip
ENTEH20N21441166C2
:
ENTEGRIS
Colour:
Black
Description:
Tray chip
Seq:
02
W×D×H:
0,810×0,802×0,03 mm
Pk:
400
Multipurpose trays for a wide range of uses.