Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Iodocumene
2-Iodocumene 95%
2-Iodocumene 95%
2-Iodocumene 95%
L01533.06
:
Thermo Fisher Scientific
:
2-Iodocumene 95%
L01533.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
246.09 g/mol
:
110 °C (20 mmHg)
:
1.54
:
242-819-8
:
GHS07
:
Ambient
:
19099-54-8
:
C₉H₁₁I
:
MFCD00013704