Deoxycholic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 12α-Hydroxylithocholic acid, (R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,12α-Dihydroxy-5β-cholanic acid, 3α,12α-Dihydroxy-5β-cholan-24-oic acid, DCA, 12alpha-Hydroxylithocholic acid, 3α,12α-Dihydroxycholan-24-oic acid, 3α,12α-Dihydroxycholanic acid
CAS Number:83-44-3
:
Formula:C₂₄H₄₀O₄
MDL Number:MFCD00003673
:392.58 g/mol
Merck Index:14,2899
Boiling Pt:504 °C (1013 hPa)
Melting Pt:172…176 °C
Storage Temperature:Ambient
EINECS:201-478-5
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together