Phenylpropiolic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:637-44-5
:
Formula:C₉H₆O₂
MDL Number:MFCD00004361
:146.15 g/mol
Boiling Pt:294 °C (1013 hPa)
Melting Pt:135…138 °C
Density:1.28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-285-8
:GHS07
Related Information
Frequently Bought Together