Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Phenyl chloroformate ≥99%

Toxic
Corrosive
Irritant
Phenyl chloroformate ≥99%
Certificates
Phenyl chloroformate ≥99%
Phenyl chloroformate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 1885-14-9 Molecular Formula: ClCO2C6H5 Formula Weight: 156.57 Storage Temperature: 2°C to 8°C Flash Point: 76°(169°F) Refractive Index: 1.5110 Melting Point: -38° Boiling Point: 188-189° Beilstein Reference: 606778 UN No.: 2746 MDL No.: MFCD00000637

  • CAS Number:1885-14-9
  • Formula:C₇H₅ClO₂
  • MDL Number:MFCD00000637
  • Molecular Weight:156.57 g/mol
  • Boiling Pt:188…189 °C
  • Melting Pt:–38 °C
  • Density:1.246
  • Flash Pt:76 °C (169 °F)
  • EINECS:217-547-8
  • UN:2746
  • ADR:6.1,II
Product Family Options

Product Information