
Tricine 99+% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5704-04-1
:
Formula:(HOCH₂)₃CNHCH₂CO₂H
MDL Number:MFCD00004277
:179.17 g/mol
Merck Index:13,09724
Melting Pt:180…184 °C (dec.)
Density:1,523 g/cm³ (19.85 °C)
EINECS:227-193-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- UV:(0.1 M in water)
- pH:4.6 to 5.6 (1 % in water at 25°C)
- Titration with HClO4:≥99.0 %
- Lead (Pb):≤5 ppm
Specification Test Results
Frequently Bought Together