

Diammonium tetrachloropalladate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13820-40-1
:
Formula:H₈Cl₄N₂Pd
MDL Number:MFCD00010884
:284.31 g/mol
Density:2.17 g/cm³
EINECS:237-498-6
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Green to red to brown Powder or crystals
- Palladium (Pd):≥36.5 %
Specification Test Results
Frequently Bought Together