Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Polyester Lab Microscope Slide Labels, Core Small

 :   Brady
Polyester Lab Microscope Slide Labels, Core Small
 :  Brady
 
Polyester Lab Microscope Slide Labels, Core Small
Polyester Lab Microscope Slide Labels, Core Small

 

 

This B-488 material combined with the R-4300 series ribbon provides a solution that can withstand various chemicals without damage, such as xylene and other chemicals commonly used in slide staining. Printing does not smear, the label does not curl, and there is no discoloration that could make barcodes illegible. The label adheres reliably to the slide.

  • The highly chemical resistant polyester (B-488) is designed for use in slide staining and other laboratory marking where excellent chemical resistance is required
  • Abrasion-resistant, resistant to chemicals, humidity-resistant
  • Resistant to low temperatures, chemical resistant, permanent adhesive, harsh environment
  • B-488 performs well in common slide staining with hematoxylin and eosin
  • Excellent resistance to various chemicals, oils and water
  • Application temperature from −80 to +130 °C

Applications are general labeling, healthcare and hospital labels, laboratory labeling.

 : Compliant with Directive 2011/65/EU and 2015/863.

Product Family Options

 

  • Description
    Model
    Material
    Printing method
    Colour
    Printer model
    Dimensions
     
     
  • Polyester lab microscope slide labels, Core inner diameter: 25,40 mm, thickness: 0,094 mm
    THT-194-488-2.5-SC
    Polyester
    Thermal transfer
    White
    BBP11
    22,86×19,05 mm
       
       
    1 * 1 Rolle
    1 * 1 Rolle - 236,00 €
    236,00 €
    1 * 1 Rolle
       236,00 €

  • Polyester lab microscope slide labels, Core inner diameter: 25,40 mm, thickness: 0,094 mm
    M511-EU-UK-LAB
    Polyester
    Thermal transfer
    White
    BBP11
    22,86×22,86 mm
       
       
    1 * 1 Rolle
    1 * 1 Rolle - 275,00 €
    275,00 €
    1 * 1 Rolle
       275,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.