Orders LinkContactUsLinkComponent
Sequential Sterilisation Wraps, Halyard
Sequential Sterilisation Wraps, Halyard
Supplier:   Halyard
Sequential Sterilisation Wraps, Halyard
Supplier:  Halyard
Certificates
Sequential Sterilisation Wraps, Halyard
4 Options Available Below
Product Details & Documents
The choice of sterile processing professionals, HALYARD sterilisation wraps, made of strong reliable SMS non-woven fabric, have been helping keep sterilised instruments and supply sets ready for use at a moment's notice. Ideal for use in individual medical and dental practices as well as major medical and surgery centers.
  • Industry standard for sterile processing teams around the world
  • Tear-, flame-, lint- and abrasion-resistant
  • Pre-vacuum steam sterilant penetration
  • Maintenance of package integrity (MPI) to meet FDA requirements
  • Ethylene oxide (EO) sterilant penetration and residuals
  • Class 1 flammability rating
  • Low linting
Halyard health offers six grades of wrap fabric from H100 to H600 to ensure precisely the right protection for your needs. Choosing is easy, the higher the number, the stronger the wrap. Halyard is also available in smaller sizes that meet the needs of smaller dental or medical offices.
POWERGUARD* technology for enhanced microbial barrier protection.
Product Family Options
Product Information
  • Dimensions
    Availability
    Price
  • 129-3799
    Sequential sterilisation wrap, H100
    Sequential sterilisation wrap, H100
    300×300 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 105,00 €
    105,00 €
    1 * 1.000 ST
       105,00 €
  • 129-3810
    Sequential sterilisation wrap, H100
    Sequential sterilisation wrap, H100
    380×380 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 151,00 €
    151,00 €
    1 * 1.000 ST
       151,00 €
  • 129-3811
    Sequential sterilisation wrap, H200
    Sequential sterilisation wrap, H200
    450×450 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 216,00 €
    216,00 €
    1 * 1.000 ST
       216,00 €
  • 129-4045
    Sequential sterilisation wrap, H100
    Sequential sterilisation wrap, H100
    910×910 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 197,00 €
    197,00 €
    1 * 300 ST
       197,00 €